Skip to Content
Merck
All Photos(1)

Key Documents

336726

Sigma-Aldrich

Ammonium thiosulfate

98%

Synonym(s):

Diammonium thiosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S2O3
CAS Number:
Molecular Weight:
148.21
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030427
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

crystals and lumps

mp

150 °C (lit.)

solubility

H2O: very soluble(lit.)
ethanol: insoluble(lit.)

density

1.679 g/mL at 25 °C (lit.)

SMILES string

N.N.OS(O)(=O)=S

InChI

1S/2H3N.H2O3S2/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

XYXNTHIYBIDHGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium thiosulfate is a non-toxic inorganic compound that is majorly used as a fertilizer and can be used as an alternative to cyanide based leaching. It has a hygroscopic nature and is soluble in water. It can be produced by the reaction of ammonia, water, and sulfur dioxide. It can be used as a photographic fixing salt.

Application

Ammonium thiosulfide can be used as a source of sulfur for the formation of cadmium sulfide thin films for electronic applications. It can also be used in the treatment of copper-gold ores for the recovery of gold.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfites, thiosulfates, and dithionites
Barbera JJ, et al.
Ullmann's Encyclopedia of Industrial Chemistry, 66(1-3), 1-21 (2000)
Adsorption of Copper from an Ammonia-Thiosulfate Media Using DOWEX 550A Ion Exchange Resin
Vargas C and Navarro P
International Journal of Nonferrous Metallurgy, 5(04), 33-33 (2016)
The treatment of copper-gold ores by ammonium thiosulfate leaching
Molleman E and Dreisinger D
Hydrometallurgy, 66(1-3), 1-21 (2002)
Electro-plating and characterisation of cadmium sulphide thin films using ammonium thiosulphate as the sulphur source
Abdul-Manaf NA, et al.
Journal of Materials Science: Materials in Electronics, 26(4), 2418-2429 (2015)
L Cassina et al.
Journal of hazardous materials, 231-232, 36-42 (2012-07-10)
Mercury-contaminated soils from a petrochemical plant in southern Italy were investigated to assess the phytoextraction efficiency of crop plants treated with the phytohormone, cytokinine (CK foliar treatment), and with the thioligand, ammonium thiosulfate (TS, soil application). Plant biomass, evapotranspiration, Hg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service