334502
Potassium hexachloropalladate(IV)
99%
Synonym(s):
Potassium palladium(IV) chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
K2PdCl6
CAS Number:
Molecular Weight:
397.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder
Recommended Products
Quality Level
Assay
99%
form
powder
density
2.738 g/mL at 25 °C (lit.)
SMILES string
[K+].[K+].Cl[Pd--](Cl)(Cl)(Cl)(Cl)Cl
InChI
1S/6ClH.2K.Pd/h6*1H;;;/q;;;;;;2*+1;+4/p-6
InChI key
OEDRXJBJMMJUSV-UHFFFAOYSA-H
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Haitham Tartor et al.
Animals : an open access journal from MDPI, 10(8) (2020-08-14)
Protocols used to collect fish skin mucus may inadvertently compromise the sampled fish or the resulting sample. Here, we used three methods (wiping, scraping, and absorption) to collect skin mucus from Atlantic salmon and compared their invasiveness on fish skin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service