Skip to Content
Merck
All Photos(3)

Key Documents

205885

Sigma-Aldrich

Palladium(II) chloride

ReagentPlus®, 99%

Synonym(s):

Dichloropalladium, Palladium dichloride, Palladous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

678-680 °C (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Palladium(II) chloride is a widely used starting material for preparing other palladium catalysts, reagents, and nanoparticles.

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
Palladium(II) chloride (PdCl2) can be used as a catalyst in the following processes:
  • PdCl2-CuCl2 catalytic system is used in the selective acetylation of 1o and 2o alkyl or aryl alcohols with vinyl acetate to prepare corresponding acetylated derivatives.
  • PdCl2–EDTA is used in the Suzuki cross-coupling reaction between aryl and heteroaryl halides with aryl(heteroaryl)boronic acids in water.
  • PdCl2-Cu(OAc)2 is used to synthesize polysubstituted pyrroles from phenethylamines via multiple deprotonations and deamination reactions.
  • PdCl2-PEG(300) is used in the Suzuki-Miyaura coupling reaction.
  • PdCl2-NaBH4 is used in the hydrogenation of carbon-carbon triple bonds to synthesize corresponding cis-alkenes in a polyethylene glycol-CH2Cl2 reaction medium.
  • PdCl2-(n-Bu)3N-NH4OOCH catalytic system is used in the synthesis of benzofurans via intramolecular Heck reaction.
For small-scale and high throughput uses, the product is also available as ChemBeads (919853)

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of benzofurans in ionic liquid by a PdCl2-catalyzed intramolecular Heck reaction
Xie X, et al.
Tetrahedron Letters, 45, 6235-6237 (2004)
Effective hydrogenation of carbon-carbon triple bonds by NaBH4/PdCl2 in polyethylene glycol/CH2Cl2: usefulness of peg in synthetic reactions
Suzuki N, et al.
Tetrahedron, 41(12), 2387-2392 (1985)
J W J Bosco et al.
Chemical communications (Cambridge, England), (9), 1116-1117 (2004-04-30)
PdCl(2) can catalyze the acetylation of primary and secondary alcohols with vinyl acetate. The reaction is selective and mild with high yields. Tertiary alcohols, phenols and amines are unaffected under these reaction conditions.
PEG (300)-PdCl2 promoted efficient and convenient Suzuki-Miyaura coupling of aryl chlorides with arylboronic acids
Yin Liang, et al.
Tetrahedron, 62(40), 9359-9364 (2006)
Pd nanocatalyst adorned on magnetic chitosan@ N-heterocyclic carbene: Eco-compatible Suzuki cross-coupling reaction
Sedghi R, et al.
Molecules (Basel), 21, 3048-3048 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service