Skip to Content
Merck
All Photos(2)

Key Documents

331716

Sigma-Aldrich

Gadolinium(III) acetylacetonate hydrate

99.9% trace metals basis

Synonym(s):

2,4-Pentanedione gadolinium(III) derivative, Gd(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd(C5H7O2)3 · xH2O
CAS Number:
Molecular Weight:
454.57 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder
solid

reaction suitability

core: gadolinium
reagent type: catalyst

mp

143 °C (dec.) (lit.)

SMILES string

O.O.CC(=O)\C=C(/C)O[Gd](O\C(C)=C\C(C)=O)O\C(C)=C\C(C)=O

InChI

1S/3C5H8O2.Gd.2H2O/c3*1-4(6)3-5(2)7;;;/h3*3,6H,1-2H3;;2*1H2/q;;;+3;;/p-3/b3*4-3+;;;

InChI key

VDRNKMRBGOSTIC-OCLMOVOUSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service