Skip to Content
Merck
All Photos(1)

Key Documents

331546

Sigma-Aldrich

2,5-Bis(trifluoromethyl)aniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3NH2
CAS Number:
Molecular Weight:
229.12
Beilstein:
2653046
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

70-71 °C/15 mmHg (lit.)

density

1.467 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Nc1cc(ccc1C(F)(F)F)C(F)(F)F

InChI

1S/C8H5F6N/c9-7(10,11)4-1-2-5(6(15)3-4)8(12,13)14/h1-3H,15H2

InChI key

XWMVIJUAZAEWIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2,5-Bis(trifluoromethyl)aniline may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumya Mukherjee et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(45), 11771-11778 (2018-05-29)
Fluorous organic building blocks were utilized to develop two self-assembled, hydrophobic, fluorinated porous organic polymers (FPOPs), namely, FPOP-100 and FPOP-101. Comprehensive mechanical analyses of these functionalised triazine network polymers marked the introduction of mechanical stiffness among all porous organic network

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service