Skip to Content
Merck
All Photos(3)

Key Documents

329436

Sigma-Aldrich

Barium carbonate

99.98% trace metals basis

Synonym(s):

Barium monocarbonate, Pigment White 10, Witherite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCO3
CAS Number:
Molecular Weight:
197.34
Beilstein:
7045119
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.98% trace metals basis

form

powder

impurities

≤250.0 ppm Trace Metal Analysis

SMILES string

[Ba++].[O-]C([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Dielectric Properties of Materials Used for Microwave-Based NOx Gas Dosimeters: This study discusses the use of barium carbonate in microwave-based gas dosimeters, highlighting its significance in material science for environmental monitoring applications (Walter et al., 2024).
  • Barium carbonate and barium titanate for ultra-high temperature thermochemical energy storage: Focuses on the use of barium carbonate in energy storage systems, particularly at ultra-high temperatures, which is crucial for advancing renewable energy technologies (Williamson et al., 2024).
  • Enhanced electrode reaction for La0. 8Sr0. 2FeO3-δ-Sm0. 2Ce0. 8O1. 9 composite cathode by barium carbonate nanoparticles as a synergistic catalyst: Discusses the catalytic properties of barium carbonate nanoparticles in solid oxide fuel cells, an important area of research in material science and sustainable energy (Li et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin V Brix et al.
Ecotoxicology and environmental safety, 73(7), 1646-1652 (2010-07-14)
A series of Toxicity Identification Evaluations (TIEs) to identify the cause(s) of observed toxicity to Ceriodaphnia dubia have been conducted on a hard rock mining effluent. Characteristic of hard rock mining discharges, the effluent has elevated (∼3000 mg l(-1)) total
Ashutosh Dash et al.
Journal of hazardous materials, 168(2-3), 1126-1133 (2009-04-03)
An attempt to recover and purify 4.66 TBq(126Ci) of (137)Cs as solution from a 20-year-old doubly encapsulated sealed source containing (137)CsCl was made successfully. The primary capsule was first cut open to retrieve the secondary capsule. The top end of
G H Kramer et al.
Health physics, 70(3), 418-424 (1996-03-01)
High level contamination was observed on two workers during a routine hand check of personnel leaving a laboratory where barium carbonate labeled with 14C was handled. A third worker was found to have been exposed to airborne 14C (carbonate) aerosol
D Ekeberg et al.
Carbohydrate research, 335(2), 141-146 (2001-09-25)
A simple, but low-yielding method for the synthesis of 3-hexuloses has been elaborated. Oxidation of 1,2:5,6-di-O-isopropylidenehexitols with bromine in the presence of barium carbonate, followed by mild-acid hydrolysis of the oxidation products gave the free hexuloses. Oxidation occurred at only
Xiaorang Tian et al.
Journal of hazardous materials, 160(2-3), 589-593 (2008-04-25)
In this study, the exploration of making acidproof fracturing proppants using red mud was carried out. The main raw materials are red mud and the refractory waste. During the exploration, three methods were explored to enhance the acid resistance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service