327611
Pentacarbonylchlororhenium(I)
98%
Synonym(s):
Rhenium(I) pentacarbonyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
98%
reaction suitability
core: rhenium
reagent type: catalyst
SMILES string
Cl[Re].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
InChI
1S/5CO.ClH.Re/c5*1-2;;/h;;;;;1H;/q;;;;;;+1/p-1
InChI key
JQUUAHKBIXPQAP-UHFFFAOYSA-M
Application
Used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands and also to prepare [3]-ferrocenophane complexes. Employed in preparation of a series of mixed-valence compounds of rhenium and iron these compounds find application in the study of charge-transfer processes.
Other Notes
For a review of organometallic complexes of rhenium, see Science of Synthesis.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sci. Synth., 2, 111-111 (2003)
Inorganic Chemistry, 10, 606-606 (1999)
ACS omega, 5(22), 12944-12954 (2020-06-18)
The Re(I) dimer complex, [fac(CO)3(phen)Re1-N(py)COORe2(phen)fac(CO)3]+ (py = pyridine; phen = 1,10-phenanthroline), contains two different Re(I) centers 9.3 Å apart, one with a nitrogen donor and the other with an acetate donor from the bridging isonicotinate ligand. The complexes were characterized
Inorganic Chemistry, 29, 253-253 (1990)
Inorganic Chemistry, 29, 2909-2909 (1990)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service