Skip to Content
Merck
All Photos(2)

Key Documents

325732

Sigma-Aldrich

Holmium(III) nitrate pentahydrate

99.9% trace metals basis

Synonym(s):

Holmium(3+) Trinitrate Pentahydrate, Nitric acid Holmium(3+) Salt Pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ho(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
441.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: holmium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.[Ho+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ho.3NO3.5H2O/c;3*2-1(3)4;;;;;/h;;;;5*1H2/q+3;3*-1;;;;;

InChI key

BJUGWWDCFYEYOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ho(NO3)3 may be used as a precursor during the synthesis of GdVO4:Ln3+/Yb3+ (Ln3+ = Er3+, Ho3+, Tm3+, Tm3+/Ho3+/Er3+) by chemical co-precipitation technique.[1] It may be used to prepare poly[[tetraaquadi-μ5-fumarato-μ4-fumaratodiholmium(III)] trihydrate].[2]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly [[tetraaquadi-?5-fumarato-?4-fumarato-diholmium (III)] trihydrate].
Rahahlia N, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(5), m1266-m1268 (2007)
Multicolor upconversion luminescence of GdVO4:Ln3+/Yb3+ (Ln3+ = Ho3+, Er3+, Tm3+, Ho3+/Er3+/Tm3+) nanorods
Gavrilovic TV, et al.
Dyes and Pigments, 126(1-7) (2016)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service