Skip to Content
Merck
All Photos(1)

Key Documents

325066

Sigma-Aldrich

(R)-(−)-3-Bromo-2-methyl-1-propanol

97%

Synonym(s):

(R)-3-Bromo-2-methylpropan-1-ol, (R)-3-Hydroxy-2-methylpropyl bromide, 3-Bromo-2-(R)-methyl-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
153.02
Beilstein:
3600269
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]25/D −6.6°, c = 2 in chloroform

refractive index

n20/D 1.484 (lit.)

bp

73-74 °C/9 mmHg (lit.)

density

1.461 g/mL at 20 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

C[C@H](CO)CBr

InChI

1S/C4H9BrO/c1-4(2-5)3-6/h4,6H,2-3H2,1H3/t4-/m0/s1

InChI key

KIBOHRIGZMLNNS-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(−)-3-Bromo-2-methyl-1-propanol is used as a starting material in the total syntheses of epothilone C and bistramide A. It can also be used as a structural modifier in the homochiral porous molecular networks.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Multi?step application of immobilized reagents and scavengers: a total synthesis of epothilone C.
Storer R I, et al.
Chemistry?A European Journal , 10(10), 2529-2547 (2004)
Control and induction of surface-confined homochiral porous molecular networks.
Tahara K, et al.
Nature Chemistry, 3(9), 714-719 (2011)
Enantioselective total synthesis of bistramide A.
Crimmins M T and DeBaillie A C
Journal of the American Chemical Society, 128(15), 4936-4937 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service