Skip to Content
Merck
All Photos(1)

Key Documents

256536

Sigma-Aldrich

Phosphorus tribromide

99%

Synonym(s):

Phosphorus(III) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PBr3
CAS Number:
Molecular Weight:
270.69
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.27 psi ( 54 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.697 (lit.)

bp

175 °C (lit.)

mp

−41.5 °C (lit.)

density

2.88 g/mL at 20 °C (lit.)

SMILES string

BrP(Br)Br

InChI

1S/Br3P/c1-4(2)3

InChI key

IPNPIHIZVLFAFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus tribromide (PBr3) is commonly used in Hell-Volhard-Zelinsky halogenation for the α-bromination of carboxylic acids to form the corresponding acyl bromide. It is also useful for the conversion of primary and secondary alcohols to alkyl bromides.

Application

Phosphorus tribromide may be used as a reagent:
  • In a novel protocol for synthesizing 7-ethoxy-1-(p-ethylphenoxy)-3,7-dimethyl-2-octene, a synthetic juvenile hormone mimic of trans,trans,cis-10,11-epoxy-7-ethyl-3,11-dimethyltrideca-2,6-dienoate.
  • To synthesize 1,2,3-diazaphosphinane, 1,2,3-thiazaphosphinine and 1,2-azaphosphole bearing a chromone ring.
  • To synthesize 1-Bromoundec-1-ene from 10-undecen-1-ol.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Chemistry (2008)
Polymerized micelle-protected platinum clusters. Preparation and application to catalyst for visible light-induced hydrogen generation.
Toshima N, et al.
J. Macromol. Sci. Chem., 25(5-7), 669-686 (1988)
Organic Chemistry (1988)
Organic Chemistry, 905-905 null
Phosphorus tribromide promoted allylic rearrangement of a tertiary vinyl carbinol. Stereochemistry of the reaction product and application to the synthesis of JH-25, a potent juvenile hormone mimic.
Babler JH
The Journal of Organic Chemistry, 41(7), 1262-1264 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service