319813
Methyl cyanoformate
99%
Synonym(s):
2-Methoxy-2-oxoacetonitrile, Cyanoformic acid methyl ester, Mander′s reagent, Methyl carbonocyanidate, [(Cyanocarbonyl)oxy]methane
About This Item
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.374 (lit.)
bp
100-101 °C (lit.)
density
1.072 g/mL at 25 °C (lit.)
functional group
ester
SMILES string
COC(=O)C#N
InChI
1S/C3H3NO2/c1-6-3(5)2-4/h1H3
InChI key
OBWFJXLKRAFEDI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
78.8 °F - closed cup
Flash Point(C)
26 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service