Skip to Content
Merck
All Photos(1)

Key Documents

226319

Sigma-Aldrich

Ethyl isocyanoacetate

95%

Synonym(s):

α-Isocyanoacetic acid ethyl ester, (Ethoxycarbonyl)methyl isonitrile, 2-Ethoxy-2-oxoethyl isocyanide, 2-Ethyloxycarbonylmethyl isocyanide, Ethoxycarbonylmethyl isocyanide, Ethyl α-isocyanoacetate, Ethyl 2-isocyanoacetate, Isocyanoacetic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CNCH2COOCH2CH3
CAS Number:
Molecular Weight:
113.11
Beilstein:
3588613
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.418 (lit.)

bp

194-196 °C (lit.)

density

1.035 g/mL at 25 °C (lit.)

functional group

amine
ester
isonitrile

storage temp.

2-8°C

SMILES string

CCOC(=O)C[N+]#[C-]

InChI

1S/C5H7NO2/c1-3-8-5(7)4-6-2/h3-4H2,1H3

InChI key

FPULFENIJDPZBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl isocyanoacetate is an isocyanide ester that serves as a building block for the production of heterocycles.

Application

Ethyl isocyanoacetate was used in the synthesis of 7-aza-tetrahydroindoles. It was also used to prepare pyrroles, oxazolines, benzodiazepines, oxazoles and imidazoles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 35, 2493-2493 (1994)
Marcus Baumann et al.
Organic letters, 8(23), 5231-5234 (2006-11-03)
[Structure: see text] A multipurpose mesofluidic flow reactor capable of producing gram quantities of material has been developed as an automated synthesis platform for the rapid on-demand synthesis of key building blocks and small exploratory libraries. The reactor is configured
Tetrahedron Letters, 34, 5463-5463 (1993)
Yifei Li et al.
Chemical communications (Cambridge, England), 48(100), 12228-12230 (2012-11-13)
A novel and efficient route for the synthesis of 7-aza-tetrahydroindoles from N-aryl/alkyl-alkenoylacetamides and ethyl isocyanoacetate is described. A mechanism, involving a stepwise [3+2] cycloaddition-intramolecular aza-Michael addition cascade, is proposed that explains the origin of the double nucleophilic attack on the
Chikashi Kanazawa et al.
Journal of the American Chemical Society, 128(33), 10662-10663 (2006-08-17)
The copper-catalyzed reaction between two different isocyanides produces the corresponding heteroaromatization products, imidazoles, in good yields. The reaction proceeds most probably through the activation of the sp3 C-H bond in the isocyanides by a copper catalyst, followed by a [3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service