138509
3-Chlorotoluene
98%
Synonym(s):
1-Chloro-3-methylbenzene, 1-Methyl-3-chlorobenzene, 3-Chloro-1-methylbenzene, 3-Methylchlorobenzene, 3-Methylphenyl chloride, m-Chlorotoluene, m-Methylchlorobenzene, m-Tolyl chloride
About This Item
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.522 (lit.)
bp
160-162 °C (lit.)
mp
−48 °C (lit.)
density
1.072 g/mL at 25 °C (lit.)
SMILES string
Cc1cccc(Cl)c1
InChI
1S/C7H7Cl/c1-6-3-2-4-7(8)5-6/h2-5H,1H3
InChI key
OSOUNOBYRMOXQQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
125.6 °F - closed cup
Flash Point(C)
52 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service