Skip to Content
Merck
All Photos(1)

Key Documents

317632

Sigma-Aldrich

(Methylcyclopentadienyl)manganese(I) tricarbonyl

Synonym(s):

(Methylcyclopentadienyl)tricarbonylmanganese, MCMT, Tricarbonyl(2-methylcyclopentadienyl)manganese, Ethyl MMT, MMT, NSC 22316

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H4CH3Mn(CO)3
CAS Number:
Molecular Weight:
218.09
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

0.05 mmHg ( 20 °C)

form

liquid

reaction suitability

core: manganese
reagent type: catalyst

refractive index

n20/D 1.584 (lit.)

bp

232-233 °C (lit.)

mp

−1 °C (lit.)

density

1.38 g/mL at 25 °C (lit.)

SMILES string

[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].C[C]1[CH][CH][CH][CH]1

InChI

1S/C6H7.3CO.Mn/c1-6-4-2-3-5-6;3*1-2;/h2-5H,1H3;;;;

InChI key

LYHJNAIHGFWRKM-UHFFFAOYSA-N

General description

Methylcyclopentadienyl manganese tricarbonyl (MMT) is commonly used as an antiknock agent in gasoline. It is also used as an additive in unleaded gasoline.

Application

Methylcyclopentadienyl manganese tricarbonyl can be used for doping of ZnSe nanocrystals.
It can also be used as a reactant for:
  • Aldol addition reactions.
  • Preparation of homo- and heteronuclear mixed biscarbene complexes with conjugated bithiophene units.
  • Molecular fragmentation using shaped femtosecond laser pulses.
  • Preparation of MnAs thin films grown on GaAs(001) by metalorganic vapour phase epitaxy (MOVPE).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Bhuie et al.
The Science of the total environment, 339(1-3), 167-178 (2005-03-03)
The environmental impacts of gasoline additives such as lead (Pb) and Methyl Tertiary Butyl Ether (MTBE) are well documented, leading to the phasing out of these additives. In contrast, little is known about the health and environmental impacts of potential
Kinjal J Shah et al.
Journal of colloid and interface science, 529, 90-99 (2018-06-11)
A variety of quaternary ammonium salts with different carbon chain lengths were used to modify and convert montmorillonite clay to organoclays. The surface modification attributing to the wettability was investigated using various techniques. The zeta-potential values of all clays showed
D R Lynam et al.
Neurotoxicology, 20(2-3), 145-150 (1999-07-01)
Methylcyclopentadienyl Manganese Tricarbonyl (MMT) has been used since the 1970s in the U.S. as a gasoline octane enhancer Extensive testing of the effects of MMT on regulated gaseous emissions carried out on a wide variety of automobiles showed that use
Minfeng Zeng et al.
ACS applied materials & interfaces, 8(48), 33157-33164 (2016-12-10)
Metal nanoparticles, once supported by a suitable scaffolding material, can be used as highly efficient heterogeneous catalysts for numerous organic reactions. The challenge, though, is to mitigate the continuous loss of metals from the supporting materials as reactions proceed, so
Vadim V Lozovoy et al.
The journal of physical chemistry. A, 112(17), 3789-3812 (2008-04-25)
The possibility that chemical reactions may be controlled by tailored femtosecond laser pulses has inspired recent studies that take advantage of their short pulse duration, comparable to intramolecular dynamics, and high peak intensity to fragment and ionize molecules. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service