Skip to Content
Merck
All Photos(4)

Key Documents

317047

Sigma-Aldrich

(−)-B-Methoxydiisopinocampheylborane

Synonym(s):

(−)-Diisopinocampheylmethoxyborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H37BO
CAS Number:
Molecular Weight:
316.33
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

SMILES string

COB([C@H]1CC2CC([C@@H]1C)C2(C)C)[C@H]3CC4CC([C@@H]3C)C4(C)C

InChI

1S/C21H37BO/c1-12-16-8-14(20(16,3)4)10-18(12)22(23-7)19-11-15-9-17(13(19)2)21(15,5)6/h12-19H,8-11H2,1-7H3/t12-,13-,14-,15-,16+,17+,18-,19-/m0/s1

InChI key

IAQXEQYLQNNXJC-BAMGFKBFSA-N

General description

B-Methoxydiisopinocampheylborane (Ipc2BOMe) is an organoborane compound, which is prepared from excess α-pinene, borane dimethylsulfide, and methanol via the formation of an intermediate diisocampheylborane. Ipc2BOMe is used as a versatile reagent for the construction of C-C bonds in asymmetric synthesis.

Application

Reactant involved in organic synthesis reactions such as:
  • Double allylboration for synthesis of fragments of tetrafibricin
  • Anticancer cytotoxic monorhizopodin synthesis
  • Annulation of cyclic allylsilanes
  • Asymmetric synthesis of β-amino-α-hydroxy acid taxol side chain analogs

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B-Methoxydiisopinocampheylborane (Ipc2BOMe): A Pinene Based Auxiliary for Asymmetric C-C Bond-Forming Reactions
Hertweck C and Boland W
J. Prakt. Chem., 341(1), 83-87 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service