317012
(+)-DIP-Chloride™
90-105% (approx.)
Synonym(s):
(+)-B-Chlorodiisopinocampheylborane, (+)-Ipc2BCl
About This Item
Recommended Products
Quality Level
Assay
90-105% (approx.)
form
solid
mp
53-55 °C (lit.)
SMILES string
C[C@@H]1[C@H](C[C@H]2C[C@@H]1C2(C)C)B(Cl)[C@H]3C[C@H]4C[C@@H]([C@@H]3C)C4(C)C
InChI
1S/C20H34BCl/c1-11-15-7-13(19(15,3)4)9-17(11)21(22)18-10-14-8-16(12(18)2)20(14,5)6/h11-18H,7-10H2,1-6H3/t11-,12-,13+,14+,15-,16-,17-,18-/m0/s1
InChI key
PSEHHVRCDVOTID-NAVXHOJHSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service