309206
Rhodium(III) nitrate solution
~10 % (w/w) (Rh in >5 wt. % HNO3)
Synonym(s):
Aqueous rhodium nitrate, Rhodium trinitrate, Rhodium(3+)trinitrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
Quality Level
concentration
≥5.0% (NaOH basis (wt% HNO3, titration))
~10 % (w/w) (Rh in >5 wt. % HNO3)
density
1.41 g/mL at 25 °C
SMILES string
[Rh]
InChI
1S/Rh
InChI key
MHOVAHRLVXNVSD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- The saga of rhodium (III) nitrate complexes and their speciation in solution: An integrated experimental and quantum chemical study of rhodium(III) nitrate complexes in solution (SN Vorobyeva et al., 2022).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of hazardous materials, 298, 282-293 (2015-06-16)
Although asbestos represents today one of the most harmful contaminant on Earth, in 72% of the countries worldwide only amphiboles are banned while controlled use of chrysotile is allowed. Uncertainty on the potential toxicity of chrysotile is due to the
Angewandte Chemie (International ed. in English), 54(46), 13715-13718 (2015-09-18)
A new type of intermolecular rhodium(II)-catalyzed [5+3] cycloaddition has been developed. This higher-order cycloaddition between pyridinium zwitterion 1,5-dipole equivalents and enol diazoacetates enables the formation of eight-membered heterocyclic skeletons, which are otherwise difficult to construct. The optimized cycloaddition occurs efficiently under
Medical physics, 42(11), 6294-6308 (2015-11-02)
Large area x-ray imagers based on complementary metal-oxide-semiconductor (CMOS) active pixel sensor (APS) technology have been proposed for various medical imaging applications including digital breast tomosynthesis (DBT). The low electronic noise (50-300 e-) of CMOS APS x-ray imagers provides a
Nutrients, 7(1), 608-624 (2015-01-20)
Selenium status of the Danish population is below that assumed optimal for the suggested protective effects against chronic diseases, including certain cancers. Fish and shellfish are important dietary sources of selenium in Denmark. We investigated the effect of increased fish
Angewandte Chemie (International ed. in English), 53(48), 13083-13087 (2014-10-01)
A regio-, diastereo-, and enantioselective [4+3] cycloaddition between vinylcarbenes and dienes has been achieved using the dirhodium tetracarboxylate catalyst [Rh2(S-BTPCP)4]. This methodology provides facile access to 1,4-cycloheptadienes that are regioisomers of those formed from the tandem cyclopropanation/Cope rearrangement reaction of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service