Merck
  • Reversal of the regiochemistry in the rhodium-catalyzed [4+3] cycloaddition between vinyldiazoacetates and dienes.
Reversal of the regiochemistry in the rhodium-catalyzed [4+3] cycloaddition between vinyldiazoacetates and dienes.
Angewandte Chemie (International ed. in English) (2014-10-01)
Pablo E Guzmán, Yajing Lian, Huw M L Davies
ABSTRACT
A regio-, diastereo-, and enantioselective [4+3] cycloaddition between vinylcarbenes and dienes has been achieved using the dirhodium tetracarboxylate catalyst [Rh2(S-BTPCP)4]. This methodology provides facile access to 1,4-cycloheptadienes that are regioisomers of those formed from the tandem cyclopropanation/Cope rearrangement reaction of vinylcarbenes with dienes.
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%
Rhodium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rhodium, microfoil, disks, 10mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%
Rhodium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%