Skip to Content
Merck
All Photos(4)

Key Documents

341010

Sigma-Aldrich

Bismuth(III) iodide

99%

Synonym(s):

Bismuth triiodide, Triiodobismuthane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiI3
CAS Number:
Molecular Weight:
589.69
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

reaction suitability

core: bismuth
reagent type: catalyst

mp

408 °C (lit.)

density

5.78 g/mL at 25 °C (lit.)

SMILES string

I[Bi](I)I

InChI

1S/Bi.3HI/h;3*1H/q+3;;;/p-3

InChI key

KOECRLKKXSXCPB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Bismuth(III) iodide or Bismuth triiodide are nontoxic and hence they are used as environmentally friendly catalysts. It can be used as a catalyst in the:
  • Chemoselective deprotection of acetals and ketals in water.
  • Synthesis of polysubstituted guanidines by guanylation reaction of thioureas with amines.
  • Sulfenylation of carbonyl compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmentally friendly organic synthesis using bismuth compounds: bismuth (III) iodide catalyzed deprotection of acetals in water
Bailey AD, et al.
Tetrahedron Letters, 49(4), 691-694 (2008)
Bismuth (III) Halides and Sulfate as Highly Efficient Catalyst for the Sulfenylation of Carbonyl and Related Compounds1
Komatsu N, et al.
Synlett, 1995(09), 984-986 (1995)
The first bismuth (III)-catalyzed guanylation of thioureas
Cunha S and Rodrigues Jr MT
Tetrahedron Letters, 47(39), 6955-6956 (2006)
Minhua Su et al.
Journal of hazardous materials, 229-230, 72-82 (2012-06-20)
BiOI-MWCNT composites, with high absorption and visible-light photocatalytic performance, were synthesized by a solvothermal process, in which ethylene glycol (EG) participated in the reaction. Synthesized BiOI-MWCNT composites were characterized by X-ray diffraction (XRD), diffuse reflectance spectra (DRS), scanning electronic microscopy
Yang Chen et al.
Physical chemistry chemical physics : PCCP, 13(13), 5659-5667 (2011-02-12)
The solvothermal reactions of BiI(3), KI, I(2), 4,4'-bipyridine (4,4'-bipy), and a small amount of water in alcohol and acetonitrile produced four bipyridinium iodobismuthates {[MQ](3)[Bi(2)I(6)(μ-I)(3)][Bi(2)I(6)(μ-I)(2)(MQ)(2)](3)} (1, MQ(+) = N-methyl-4,4'-bipyridinium), {[EQ](3)[Bi(2)I(6)(μ-I)(3)][Bi(2)I(6)(μ-I)(2)(EQ)(2)](3)} (2, EQ(+) = N-ethyl-4,4'-bipyridinium), [MV][BiI(5)] (Eur. J. Inorg. Chem., 2010, 5326)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service