Skip to Content
Merck
All Photos(1)

Key Documents

306932

Sigma-Aldrich

3,4-Hexanedione

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CH2COCOCH2CH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
1700837
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.41 (lit.)

bp

131 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCC(=O)C(=O)CC

InChI

1S/C6H10O2/c1-3-5(7)6(8)4-2/h3-4H2,1-2H3

InChI key

KVFQMAZOBTXCAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The dermal irritation and sensitization potential of 3,4-hexanedione (used as constituents of synthetic flavoring agent) was studied using a murine model. The 3,4-hexanedione derivatives inhibited oxoglutarate transport.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Campillo et al.
Bioanalysis, 9(4), 369-379 (2017-01-20)
Two important markers of oxidative stress, glyoxal and methylglyoxal, are preconcentrated from human urine by surfactant-assisted dispersive liquid-liquid microextraction and separated by LC-fluorescence. Derivatization was carried out overnight with 0.8 mM 2,3-diaminonaphthalene at 4°C. For surfactant-assisted dispersive liquid-liquid microextraction, 500
Stacey E Anderson et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 62, 373-381 (2013-09-07)
Concern has been raised over the association of diacetyl with lung disease clinically resembling bronchiolitis obliterans in food manufacturing workers. This has resulted in the need for identification of alternative chemicals to be used in the manufacturing process. Structurally similar
Elizabeth I Parkinson et al.
Bioorganic & medicinal chemistry, 19(15), 4635-4643 (2011-07-08)
Carboxylesterases (CE) are ubiquitous enzymes found in both human and animal tissues and are responsible for the metabolism of xenobiotics. This includes numerous natural products, as well as a many clinically used drugs. Hence, the activity of these agents is
I Stipani et al.
Archives of biochemistry and biophysics, 331(1), 48-54 (1996-07-01)
The effect of arginine-specific reagents on the function of the purified and reconstituted oxoglutarate carrier protein of the inner mitochondrial membrane has been investigated. The alpha-dicarbonyl reagents 2,3-butanedione, 2,3-pentanedione, 2,3- and 3,4-hexanedione, 1-phenyl-1,2-propanedione, phenylglyoxal, and phenylglyoxal derivatives caused a concentration-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service