Skip to Content
Merck
3,4-Hexanedione 95%

3,4-Hexanedione

3,4-Hexanedione 95%
Linear Formula:
CH2CH2COCOCH2CH3
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing