306878
2-Bromo-2-methylbutane
95%
Synonym(s):
tert-Amyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3CH2CBr(CH3)2
CAS Number:
Molecular Weight:
151.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.4423 (lit.)
bp
107 °C/735 mmHg (lit.)
density
1.182 g/mL at 25 °C (lit.)
functional group
alkyl halide
bromo
SMILES string
CCC(C)(C)Br
InChI
1S/C5H11Br/c1-4-5(2,3)6/h4H2,1-3H3
InChI key
JOUWCKCVTDSMHF-UHFFFAOYSA-N
General description
The enthalpies of the solution of 2-bromo-2-methylbutane and 2-bromo-2-methylpropane in mono- and dialcohols were studied.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
41.0 °F - closed cup
Flash Point(C)
5 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The enthalpies of solution of 2-bromo-2-methylpropane and 2-bromo-2-methylbutane in mono-and dialcohols at 298.15 K.
Albuquerque LM, et al.
Thermochimica Acta, 322(1), 57-61 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service