Skip to Content
Merck
All Photos(3)

Key Documents

124095

Sigma-Aldrich

1-Bromo-3-methylbutane

96%

Synonym(s):

Isoamyl bromide, Isopentyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2Br
CAS Number:
Molecular Weight:
151.04
Beilstein:
773652
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.4409 (lit.)

bp

120-121 °C (lit.)

mp

−112 °C (lit.)

solubility

H2O: slightly soluble at 16.5 °C (0.02g/100ml)
alcohol: miscible
diethyl ether: miscible

density

1.261 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

storage temp.

2-8°C

SMILES string

CC(C)CCBr

InChI

1S/C5H11Br/c1-5(2)3-4-6/h5H,3-4H2,1-2H3

InChI key

YXZFFTJAHVMMLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-3-methylbutane is a low toxic bromosolvent.

Application

1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole. It was used in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mei-I Leong et al.
Journal of chromatography. A, 1217(34), 5455-5461 (2010-07-29)
A low toxic dispersive liquid-liquid microextraction (LT-DLLME) combined with gas chromatography-mass spectrometry (GC-MS) had been developed for the extraction and determination of 16 polycyclic aromatic hydrocarbons (PAHs) in water samples. In normal DLLME assay, chlorosolvent had been widely used as
Erin N Sharp et al.
The journal of physical chemistry. A, 112(7), 1445-1456 (2008-01-24)
Cavity ringdown spectra of the A-X electronic transition of all eight isomers of the pentyl peroxy radical are reported. Using the corresponding assignments from previously studied smaller alkyl peroxy radicals, assignments of origin bands are made for the pentyl peroxy
J Stephen Elmore et al.
Journal of agricultural and food chemistry, 56(15), 6173-6179 (2008-07-16)
Wheat flour from plants deficient in sulfur has been shown to contain substantially higher levels of free amino acids, particularly asparagine and glutamine, than flour from wheat grown where sulfur nutrition was sufficient. Elevated levels of asparagine resulted in acrylamide
Guilong Peng et al.
Archives of environmental contamination and toxicology, 70(3), 607-614 (2015-11-29)
A low toxic solvent-based vortex-assisted surfactant-enhanced emulsification liquid-liquid microextraction (LT-VSLLME) combined with graphite furnace atomic absorption spectrometry was developed for the extraction and determination of lead (Pb) in water samples. In the LT-VSLLME method, the extraction solvent was dispersed into
Peter Tieu et al.
Frontiers in chemistry, 8, 599-599 (2020-08-06)
Metal nanoparticle catalysts functionalized with small, well-defined organic ligands are important because such systems can provide a spatial control in the catalyst-substrate interactions. This article describes the synthesis, stability, and catalytic property evaluations of four different Pd nanoparticles capped with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service