Skip to Content
Merck
All Photos(1)

Key Documents

306657

Sigma-Aldrich

Heptadecanoyl chloride

98%

Synonym(s):

Margaroyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15COCl
CAS Number:
Molecular Weight:
288.90
Beilstein:
1781006
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

176 °C/4 mmHg (lit.)

density

0.883 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCC(Cl)=O

InChI

1S/C17H33ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3

InChI key

ICDQUAGMQCUEMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptadecanoyl chloride reacts with 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine to form 1-palmitoyl-2-oleoyl-N-heptadecanoyl phosphatidylethanolamine.

Application

Heptadecanoyl chloride was used in the synthesis of N-heptadecanoylethanolamine by reacting with ethanolamine.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabiana Piscitelli et al.
Nutrition & metabolism, 8(1), 51-51 (2011-07-14)
Omega-3 polyunsaturated fatty acids (ω-3-PUFA) are known to ameliorate several metabolic risk factors for cardiovascular disease, and an association between elevated peripheral levels of endogenous ligands of cannabinoid receptors (endocannabinoids) and the metabolic syndrome has been reported. We investigated the
Saori Oka et al.
The Journal of biological chemistry, 280(18), 18488-18497 (2005-03-08)
2-Arachidonoylglycerol is an endogenous ligand for the cannabinoid receptors. Two types of cannabinoid receptors have been identified to date. The CB1 receptor is abundantly expressed in the brain, and assumed to be involved in the attenuation of neurotransmission. On the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service