306258
Poly(4-vinylpyridine-co-butyl methacrylate)
~90% 4-vinylpyridine
About This Item
Recommended Products
form
solid
Quality Level
quality
~90% 4-vinylpyridine
SMILES string
n1ccc(cc1)C=C.O(CCCC)C(=O)C(=C)C
InChI
1S/C8H14O2.C7H7N/c1-4-5-6-10-8(9)7(2)3;1-2-7-3-5-8-6-4-7/h2,4-6H2,1,3H3;2-6H,1H2
InChI key
CBBZCNWNEFEHRW-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.
Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.
Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.
Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service