Skip to Content
Merck
All Photos(2)

Key Documents

192074

Sigma-Aldrich

Poly(4-vinylpyridine-co-styrene)

powder

Synonym(s):

4-Vinylpyridine-styrene copolymer, Poly(4-vinylpyridine)-polystyrene copolymer, Styrene-4-vinylpyridine copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

application(s)

advanced drug delivery
coatings

SMILES string

C=CC1=CC=CC=C1.C=CC1=CC=NC=C1

InChI

1S/C8H8.C7H7N/c1-2-8-6-4-3-5-7-8;1-2-7-3-5-8-6-4-7/h2-7H,1H2;2-6H,1H2

InChI key

CIPXQVPZUWMSQE-UHFFFAOYSA-N

Application

Poly(4-vinylpyridine-co-styrene) can be used:
  • As a precursor in the preparation of ionic polymer with pendant photochromic moieties by reactingwith1,3,3-trimethyl-6′-bromohexyloxyspiro[2H]-indol-2,3′-[3H]-naphth[2,1-b][1,4]oxazine.
  • As permselective membrane in the preparation of composite electrodes for glucose sensing.
  • To prepare mixed redox polymer [RuIII(edta)(OH2)(py)]2[RuII(NH3)5(py)], which can switch into six redox states.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yeong-Soon Gal et al.
Journal of nanoscience and nanotechnology, 8(9), 4885-4888 (2008-12-04)
An ionic polymer with pendant photochromic moieties was prepared by the reaction of poly(4-vinylpyridine-co-styrene) and 1,3,3-trimethyl-6'-bromohexyloxyspiro[2H]-indol-2,3'-[3H]-naphth[2,1-b][1,4]oxazine. The photochromic reaction in question is caused by the reversible heterolytic cleavage of the C(spiro)-O bond under UV irradiation, yielding the colored form that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service