Skip to Content
Merck
All Photos(1)

Key Documents

303356

Sigma-Aldrich

Methyl 12-oxooctadecanoate

98%

Synonym(s):

Methyl 12-ketostearate, Methyl 12-oxostearate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CO(CH2)10CO2CH3
CAS Number:
Molecular Weight:
312.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

178-180 °C/0.8 mmHg (lit.)

mp

46-48 °C (lit.)

functional group

ester

SMILES string

CCCCCCC(=O)CCCCCCCCCCC(=O)OC

InChI

1S/C19H36O3/c1-3-4-5-12-15-18(20)16-13-10-8-6-7-9-11-14-17-19(21)22-2/h3-17H2,1-2H3

InChI key

XVSPEBNRFAFNAV-UHFFFAOYSA-N

General description

Methyl 12-oxooctadecanoate is a long chain keto fatty acid and its reaction with hydrazoic acid was studied.

Application

Methyl 12-oxooctadecanoate was used in preparation of methyl hexahydro-3-hexyl-6-thioxo-1,2,4,5-tetrazine-3-undecanoate, hexahydrothioxotetrazine fatty derivative, via reaction with thiocarbohydrazide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derivatization of keto fatty acids: Part XIII-Synthesis of methyl hexahydro-3-alkyl-6-thioxo-1, 2, 4, 5-tetrazine-3-alkanoates.
Saeed MT, et al.
Journal of the American Oil Chemists' Society, 69(4), 396-397 (1992)
Derivatization of Keto Fatty Acids IV Reaction of Hydrazoic Acid on Long Chain Keto Acid Esters.
Ahmad F, et al.
European Journal of Lipid Science and Technology, 88(2), 62-65 (1986)
Michal Korinek et al.
Frontiers in pharmacology, 8, 356-356 (2017-07-05)
Increasing prevalence of allergic diseases with an inadequate variety of treatment drives forward search for new alternative drugs. Fatty acids, abundant in nature, are regarded as important bioactive compounds and powerful nutrients playing an important role in lipid homeostasis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service