Skip to Content
Merck
All Photos(1)

Key Documents

303038

Sigma-Aldrich

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H36ClFeN4O4
CAS Number:
Molecular Weight:
824.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

λmax

421 nm

SMILES string

COc1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(OC)cc4)c5ccc6c(-c7ccc(OC)cc7)c8ccc(n8)c(-c9ccc(OC)cc9)c%10ccc2n%10[Fe](Cl)n56

InChI

1S/C48H36N4O4.ClH.Fe/c1-53-33-13-5-29(6-14-33)45-37-21-23-39(49-37)46(30-7-15-34(54-2)16-8-30)41-25-27-43(51-41)48(32-11-19-36(56-4)20-12-32)44-28-26-42(52-44)47(40-24-22-38(45)50-40)31-9-17-35(55-3)18-10-31;;/h5-28H,1-4H3;1H;/q-2;;+3/p-1/b45-37-,45-38-,46-39-,46-41-,47-40-,47-42-,48-43-,48-44-;;

InChI key

ZZRDTBXKIGUTNH-HTMHXADGSA-M

General description

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride (FeTMPPCl) is a metalloporphyrin.

Application

FeTMPPCI may be used to form interfacial molecular assemblies of metalloporphyrins. It has been used in the delignification of wood chips and pulps.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delignification of Wood Chips and Pulps by Using Natural and
Synthetic Porphyrins: Models of Fungal
Paszynski A, et al.
Applied and Environmental Microbiology, 62-68 (1998)
Effect of Heat Treatment on the Redox Properties of Iron Porphyrins Adsorbed on High Area Carbon in Acid Electrolytes:? An in Situ Fe K-Edge X-ray Absorption Near-Edge Structure Study
The Journal of Physical Chemistry B, 102 (21), 4114-4117 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service