Skip to Content
Merck
All Photos(2)

Key Documents

89456

Supelco

4,4′,4″,4″′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid)

for spectrophotometric det. of transition metals, ≥95.0%

Synonym(s):

meso-Tetraphenylporphine-4,4′,4″,4″′-tetrasulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H30N4O12S4
CAS Number:
Molecular Weight:
934.99
Beilstein:
4227566
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥95.0% (HPLC)
≥95.0%

form

powder

quality

for spectrophotometric det. of transition metals

technique(s)

UV/Vis spectroscopy: suitable

impurities

~9% water

SMILES string

OS(=O)(=O)c1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(cc4)S(O)(=O)=O)c5ccc([nH]5)c(-c6ccc(cc6)S(O)(=O)=O)c7ccc(n7)c(-c8ccc(cc8)S(O)(=O)=O)c9ccc2[nH]9

InChI

1S/C44H30N4O12S4/c49-61(50,51)29-9-1-25(2-10-29)41-33-17-19-35(45-33)42(26-3-11-30(12-4-26)62(52,53)54)37-21-23-39(47-37)44(28-7-15-32(16-8-28)64(58,59)60)40-24-22-38(48-40)43(36-20-18-34(41)46-36)27-5-13-31(14-6-27)63(55,56)57/h1-24,45,48H,(H,49,50,51)(H,52,53,54)(H,55,56,57)(H,58,59,60)/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-

InChI key

PBHVCRIXMXQXPD-LWQDQPMZSA-N

Application

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) [tetrasodium salt hydrate, TPPS] has been used in preparing Co–Al LDH/Fe(III)-TPPS film modified electrode; for demonstrating reversible redox peak attribution to Co(III)/Co(II), with iron(III) porphyrin; making it mediator for electron transfer.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

max. Absorption (in Methanol): 412-416 nm

Other Notes

Reagent for the spectrophotometric determination of ultra trace amounts of transition metals; determination by capillary zone electrophoresis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Angela Castriciano et al.
Chemical communications (Cambridge, England), 48(40), 4872-4874 (2012-04-14)
The kinetics of growth for chiral J-aggregates of H(4)TPPS(4) porphyrin have been investigated under different experimental conditions in the presence of tartaric acid. The observed rate constants and the anisotropy factor g show a defined dependence on the enantiomer used
Zsolt Valicsek et al.
Dalton transactions (Cambridge, England : 2003), 41(42), 13120-13131 (2012-09-25)
Lanthanum(III) ions form kinetically labile complexes with the 5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrin anion (H(2)TSPP(4-)), the compositions and formation constants of which significantly depend on the presence of potential axial ligands (at 0.01 M). Deviating from the chloride ion, acetate coordinating to the metal
H. Ishii, H. Tsuchiai
Analytical Sciences, 3, 229-229 (1987)
Chan Lee et al.
Toxicology, 283(2-3), 118-128 (2011-03-15)
Parkinson's disease (PD) is a progressive neurodegenerative movement disorder characterized by selective loss of dopaminergic neurons in the substantia nigra. 6-Hydroxydopamine (6-OHDA) is a catecholaminergic neurotoxin widely used to produce experimental models of PD and has been reported to cause
Christopher C Rich et al.
Physical chemistry chemical physics : PCCP, 14(7), 2362-2374 (2012-01-14)
Helical porphyrin nanotubes of tetrakis(4-sulfonatophenyl)porphyrin (TSPP) were examined in DCl/D(2)O solution using resonance Raman and resonance light scattering spectroscopy to probe the influence of hydrogen bonding on the excitonic states. Atomic force microscopy reveals similar morphology for aggregates deposited from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service