Skip to Content
Merck
All Photos(1)

Key Documents

190020

Sigma-Aldrich

Sodium cyanoborodeuteride

97 atom % D, ≥96% (CP)

Synonym(s):

Deuterated sodium borocyanohydride, Deuterated sodium cyanoborohydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBD3CN
CAS Number:
Molecular Weight:
65.86
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

97 atom % D

Assay

≥96% (CP)

form

powder

mp

>242 °C (dec.) (lit.)

mass shift

M+3

SMILES string

[Na+].[2H][B-]([2H])([2H])C#N

InChI

1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1/i2D3;

InChI key

CVDUGUOQTVTBJH-MUTAZJQDSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaoping Qing et al.
European journal of immunology, 46(12), 2737-2748 (2016-09-08)
CSF1R (colony stimulating factor 1 receptor) is the main receptor for CSF1 and has crucial roles in regulating myelopoeisis. CSF1R can be proteolytically released from the cell surface by ADAM17 (A disintegrin and metalloprotease 17). Here, we identified CSF1R as
Hucong Jiang et al.
Journal of proteome research, 18(1), 469-477 (2018-10-23)
Absolute quantification of clinical biomarkers by mass spectrometry (MS) has been challenged due to low sample-throughput of current multiple reaction monitoring (MRM) methods. For this problem to be overcome, in this work, a novel high-sample-throughput multiple reaction monitoring mass spectrometric
Yuewei Sheng et al.
Journal of the American Society for Mass Spectrometry, 30(2), 218-226 (2018-10-18)
Solvent-accessibility change plays a critical role in protein misfolding and aggregation, the culprit for several neurodegenerative diseases, including amyotrophic lateral sclerosis (ALS). Mass spectrometry-based hydroxyl radical (·OH) protein footprinting has evolved as a powerful and fast tool in elucidating protein
The Trung Tran et al.
The Analyst, 143(18), 4359-4365 (2018-08-17)
Absolute targeted proteomics typically employs known amounts of synthetic stable isotopically labeled peptides which are mixed with the analyte and analysed by LC-MS to determine the concentration of proteins. In order to obtain more data, we evaluated the use of
Alejandro Gomez-Auli et al.
Biochimie, 122, 88-98 (2015-10-13)
Carcinomas establish a molecular cross talk between malignant tumor cells and the activated non-malignant cells of the tumor stroma. This cell-cell communication in tumor-stroma interaction includes soluble, secreted proteins that act in a paracrine or autocrine manner. Proteases are crucial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service