Skip to Content
Merck
All Photos(1)

Key Documents

296163

Sigma-Aldrich

Dichloromethane-d2

≥99.5 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Dideuteromethylenechloride, Methylene chloride-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2Cl2
CAS Number:
Molecular Weight:
86.94
Beilstein:
1733318
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

24.96 psi ( 55 °C)
7.11 psi ( 20 °C)

Quality Level

isotopic purity

≥99.5 atom % D

Assay

≥99.00% (GC)

form

liquid

contains

0.03 % (v/v) TMS

packaging

ampule of 1g, 10g
glass bottle of 25g

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.422 (lit.)

bp

40 °C (lit.)

mp

-97 °C (lit.)

density

1.362 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(Cl)Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1D2

InChI key

YMWUJEATGCHHMB-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane-d2 is a deuterated derivative of dichloromethane that can be used as a solvent in the NMR spectral analysis of polymers.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mobility Studies on Siloxane-Based Inorganic-Organic Hybrid Polymers by Dynamic Solid State, Suspended State, and Deuterium NMR Spectroscopy: Supported Organometallic Complexes. XXVI [1]
Lindner E, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 1124-1131, 628-628 (2002)
Applications and new developments of the direct exponential curve resolution algorithm (DECRA). Examples of spectra and magnetic resonance images.
Windig W, et al.
Journal of Chemometrics, 13(2), 95-110 (1999)
Ting Li et al.
Environmental science and pollution research international, 22(15), 11792-11800 (2015-04-11)
We surveyed the occurrence of nine N-nitrosamine species in ten bottled drinking waters from supermarket and other water samples including raw waters, finished waters, and distribution system waters from nine municipal drinking water treatment plants in eight cities of Jiangsu
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service