Recommended Products
Quality Level
Assay
98%
form
solid
mp
98-102 °C (lit.)
functional group
bromo
carboxylic acid
SMILES string
OC(=O)Cc1cccc(Br)c1
InChI
1S/C8H7BrO2/c9-7-3-1-2-6(4-7)5-8(10)11/h1-4H,5H2,(H,10,11)
InChI key
KYNNBXCGXUOREX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
3-Bromophenylacetic acid was used in the preparation of 3-bromophenylacetaldehyde by di-isobutyl aluminum hydride (DIBAL) reduction.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of organic chemistry, 70(18), 7305-7316 (2005-08-27)
[structure: see text] Various approaches to the indole bis-oxazole fragment of the marine secondary metabolite diazonamide A are described, all of which feature dirhodium(II)-catalyzed reactions of diazocarbonyl compounds in key steps. Thus, 3-bromophenylacetaldehyde is converted into an alpha-diazo-beta-ketoester, dirhodium(II)-catalyzed reaction
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service