Skip to Content
Merck
All Photos(1)

Key Documents

260061

Sigma-Aldrich

2-Bromophenylacetic acid

99%

Synonym(s):

2-(2-Bromophenyl)acetic acid, 2-Bromobenzeneacetic acid, o-Bromophenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CO2H
CAS Number:
Molecular Weight:
215.04
Beilstein:
2250655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

104-106 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

OC(=O)Cc1ccccc1Br

InChI

1S/C8H7BrO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

DWXSYDKEWORWBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromophenylacetic acid has been used:
  • as starting reagent in the synthesis of 8-methoxy-2-tetralone
  • in the preparation of di-and tri-substituted 2-oxopiperazines on solid support

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A peptoid based synthesis of di-and tri-substituted 2-oxopiperazines on solid support.
Goff DA.
Tetrahedron Letters, 39(12), 1473-1476 (1998)
Lee S, et al.
Synthetic Communications, 25(18), 2775-2780 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service