Skip to Content
Merck
All Photos(1)

Key Documents

288101

Sigma-Aldrich

(Acetylacetonato)dicarbonylrhodium(I)

98%

Synonym(s):

Dicarbonyl-acetylacetonato-rhodium(I), Dicarbonylrhodium(I) 2,4-pentanedionate, Rh(CO)2acac, Rhodium(I) dicarbonyl acetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh(CO)2(C5H7O2)
CAS Number:
Molecular Weight:
258.03
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

reaction suitability

reagent type: catalyst

mp

154-156 °C (lit.)

SMILES string

[C-]#[O+].[C-]#[O+].CC(=O)\C=C(\C)O[Rh]

InChI

1S/C5H8O2.2CO.Rh/c1-4(6)3-5(2)7;2*1-2;/h3,6H,1-2H3;;;/q;;;+1/p-1/b4-3-;;;

InChI key

BZCAWKOPWNIDOC-FGSKAQBVSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Employed in in situ formation of a fluorous-soluble hydroformylation catalyst of interest in molecular engineering.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Horvath, I.T. et al.
Journal of the American Chemical Society, 120, 3133-3133 (1998)
Andrei V Pakoulev et al.
The journal of physical chemistry. A, 112(28), 6320-6329 (2008-06-25)
Triply vibrationally enhanced four-wave mixing spectroscopy is employed to observe vibrational coherence transfer between the asymmetric and symmetric CO-stretching modes of rhodium(I) dicarbonyl acetylacetonate (RDC). Coherence transfer is a nonradiative transition of a coherent superposition of quantum states to a
K A Merchant et al.
Physical review letters, 86(17), 3899-3902 (2001-05-01)
2D spectrally resolved ultrafast (<200 fs) IR vibrational echo experiments were performed on Rh(CO)(2)acac [(acetylacetonato)dicarbonylrhodium (I)]. The 2D spectra display features that reflect the 0-1 and 1-2 transitions and the combination band transition of the symmetric (S) and antisymmetric (A)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service