
(Acetylacetonato)dicarbonylrhodium(I)

Synonym(s):
Dicarbonyl-acetylacetonato-rhodium(I), Dicarbonylrhodium(I) 2,4-pentanedionate, Rh(CO)2acac, Rhodium(I) dicarbonyl acetylacetonate
Linear Formula:
Rh(CO)2(C5H7O2)
Products
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing