Skip to Content
Merck
All Photos(1)

Key Documents

28440

Sigma-Aldrich

tert-Butyl cyanoacetate

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2COOC(CH3)3
CAS Number:
Molecular Weight:
141.17
Beilstein:
1755933
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

refractive index

n20/D 1.420

bp

40-42 °C/0.1 mmHg (lit.)

density

0.988 g/mL at 20 °C (lit.)

functional group

ester
nitrile

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)CC#N

InChI

1S/C7H11NO2/c1-7(2,3)10-6(9)4-5-8/h4H2,1-3H3

InChI key

BFNYNEMRWHFIMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl cyanoacetate undergoes functionalization and decarboxylation to form 3-amino-4-alkyl isoquinolines.

Application

tert-Butyl cyanoacetate was used in the synthesis of vinylogous urea.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry B Snider et al.
Organic letters, 7(20), 4519-4522 (2005-09-24)
[reaction: see text] Addition of the enolate of tert-butyl acetate to cyanamide methyl ester 17 followed by treatment with LHMDS afforded vinylogous urea 19 in 27% yield. Vinylogous urea 19 was also obtained from 37 and tert-butyl cyanoacetate in 50%
Ben S Pilgrim et al.
Organic letters, 15(24), 6190-6193 (2013-11-21)
A methyl ketone, an aryl bromide, an electrophile, and ammonium chloride were combined in a four-component, three-step, and one-pot coupling procedure to furnish substituted isoquinolines in overall yields of up to 80%. This protocol utilizes the palladium catalyzed α-arylation reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service