Skip to Content
Merck
All Photos(1)

Key Documents

277363

Sigma-Aldrich

4,5-Dichloroimidazole

≥98%

Synonym(s):

4,5-Dichloro-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2Cl2N2
CAS Number:
Molecular Weight:
136.97
Beilstein:
606155
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

mp

183-185 °C (lit.)

functional group

chloro

SMILES string

Clc1nc[nH]c1Cl

InChI

1S/C3H2Cl2N2/c4-2-3(5)7-1-6-2/h1H,(H,6,7)

InChI key

QAJJXHRQPLATMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,5-Dichloroimidazole has been used in the synthesis of:
  • phosphinite complexes
  • 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Zhou et al.
Langmuir : the ACS journal of surfaces and colloids, 35(9), 3248-3255 (2019-02-15)
The detection of trace amount of volatile organic compounds (VOCs) has been covered by tons of researches, which are dedicated to improve the detection limit and insensitivity to humidity. In this work, we have synthesized ZnO@ZIF-71 nanorod arrays (NRAs) equipped
Domenico Iacopetta et al.
ChemMedChem, 12(24), 2054-2065 (2017-11-10)
Cisplatin and other platinum-based drugs are well-known valid anticancer drugs. However, during chemotherapy, the presence of numerous side effects and the onset of frequent phenomena of resistance has pushed many research groups to devise new metal-based compounds holding improved anticancer
Wai Fen Yong et al.
ChemSusChem, 10(20), 4046-4055 (2017-08-24)
Polymers containing ethylene oxide (EO) groups have gained significant interest as the EO groups have favorable interactions with polar molecules such as H
Iridium catalysts with bicyclic pyridine-phosphinite ligands: asymmetric hydrogenation of olefins and furan derivatives.
Stefan Kaiser et al.
Angewandte Chemie (International ed. in English), 45(31), 5194-5197 (2006-07-11)
Khadijah M Hindi et al.
Biomaterials, 30(22), 3771-3779 (2009-04-28)
The pressing need to treat multi-drug resistant bacteria in the chronically infected lungs of cystic fibrosis (CF) patients has given rise to novel nebulized antimicrobials. We have synthesized a silver-carbene complex (SCC10) active against a variety of bacterial strains associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service