Skip to Content
Merck
All Photos(1)

Key Documents

272078

Sigma-Aldrich

(1S)-(+)-Camphorquinone

99%

Synonym(s):

(1S)-(+)-Bornanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
Beilstein:
2613999
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]20/D +100°, c = 1.9 in toluene

mp

197-201 °C (lit.)

functional group

ketone

SMILES string

CC1(C)[C@H]2CC[C@]1(C)C(=O)C2=O

InChI

1S/C10H14O2/c1-9(2)6-4-5-10(9,3)8(12)7(6)11/h6H,4-5H2,1-3H3/t6-,10+/m0/s1

InChI key

VNQXSTWCDUXYEZ-QUBYGPBYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bioreduction of quinones and other ketones by red algae.
Chiral starting material.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Hee Shin et al.
Photomedicine and laser surgery, 29(8), 545-550 (2011-03-23)
This study evaluated the effectiveness of the diode-pumped solid state (DPSS) laser as a light source for light-curing dental resin composites. A DPSS laser of 473  nm may be useful because of its match with the absorption peak of camphorquinone
Maria Rosaria di Nunzio et al.
The journal of physical chemistry. A, 113(34), 9424-9433 (2009-08-07)
In this article, we report a study on the singlet and triplet excited-state properties of a spirooxazine (1,3-dihydro-3,3-dimethyl-1-isobutyl-6'-(2,3-dihydro-1H-indol-1-yl)spiro[2H-indole-2,3'-3H-naphtho[2,1-b][1,4]oxazine]). The singlet state of this molecule is photoreactive: upon UV light stimulation, it produces a colored merocyanine that thermally reverts to the
Ivonne Lammers et al.
Analytical chemistry, 82(22), 9410-9417 (2010-10-23)
The sensitivity of enantioselective cyclodextrin-induced room-temperature phosphorescence detection of camphorquinone (CQ) is enhanced using sensitization via a donor with a high extinction coefficient. The enantiomeric distinction is based on the different phosphorescence lifetimes of (+)-CQ and (-)-CQ after their complexation
Emerson H Silva et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(6), 790-794 (2012-04-10)
The purpose of this paper was to evaluate the influence of different light curing units on the conversion of four composite resins with different compositions (Durafill VS - Heraeus-Kulzer, Tetric Ceram - Ivoclar/Vivadent, Filtek Supreme XT - 3M ESPE e
Vesna Miletic et al.
Journal of dentistry, 40(2), 106-113 (2011-11-19)
To determine the degree of conversion (DC) over 48 h post-curing of resin mixtures containing trimethylbenzoyl-diphenylphosphine oxide (TPO) initiator cured by a polywave or a monowave LED light-curing unit (LCU). In resin mixtures based on equal weight percent (wt%) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service