Skip to Content
Merck
All Photos(1)

Key Documents

270083

Sigma-Aldrich

9-(Methylaminomethyl)anthracene

99%

Synonym(s):

(Anthracen-9-ylmethyl)-N-methylamine, 1-Anthracen-9-yl-N-methylmethanamine, 9-(N-Methylaminomethyl)anthracene, N-Methyl-9-anthracenemethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N
CAS Number:
Molecular Weight:
221.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

mp

57-59 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CNCc1c2ccccc2cc3ccccc13

InChI

1S/C16H15N/c1-17-11-16-14-8-4-2-6-12(14)10-13-7-3-5-9-15(13)16/h2-10,17H,11H2,1H3

InChI key

WRVHTDZJMNUGQN-UHFFFAOYSA-N

General description

9-(Methylaminomethyl)anthracene is an isocyanate derivatizing reagent[1].

Application

9-(Methylaminomethyl)anthracene (MAMA) was used as a model drug in the study of the effect of enzymatic degradation on the release behaviors of MAMA by UV-Vis spectroscopy[2]. It was also used as a reagent in a sensitive method for determination of isocyanates in air by HPLC via UV or fluorescence detection[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Liquid Chromatography, 3, 971-971 (1980)
X Y Xiong et al.
Journal of controlled release : official journal of the Controlled Release Society, 108(2-3), 263-270 (2005-10-04)
Poly(lactic acid) (PLA) was successfully grafted to both ends of Pluronic F127 block copolymer (PEO-PPO-PEO) to obtain amphiphilic PLA-F127-PLA block copolymers. The effect of enzymatic degradation on the release behaviors of hydrophobic model drug 9-(methylaminomethyl)anthracene (MAMA) from PLA-F127-PLA nano-particles with
R P Streicher et al.
American Industrial Hygiene Association journal, 57(10), 905-913 (1996-10-01)
Analytical reference standards generally are not available for non-monomeric isocyanate species, making accurate identification and quantitation by high-performance liquid chromatography (HPLC) difficult. A successful derivatizing reagent must react rapidly with all isocyanate groups, the derivatized isocyanate must be detectable selectively

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service