Skip to Content
Merck
All Photos(1)

Key Documents

266701

Sigma-Aldrich

Cobalt

wire, diam. 1.0 mm, 99.995% trace metals basis

Synonym(s):

Cobalt element, Cobalt-59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.995% trace metals basis

form

wire

resistivity

6.24 μΩ-cm, 20°C

diam.

1.0 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

1.4 g = 20 cm

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Perconti et al.
Journal of biological regulators and homeostatic agents, 27(2), 443-454 (2013-07-09)
Size-dependent characteristics of novel engineered nanomaterials might result in unforeseen biological responses and toxicity. To address this issue, we used cDNA microarray analysis (13443 genes) coupled with bioinformatics and functional gene annotation studies to investigate the transcriptional profiles of Balb/3T3
Dilemmas about the toxicological consequences of metal-on-metal hip prostheses -- What we do and do not know, and what we should do?
Jeffrey Brent et al.
Clinical toxicology (Philadelphia, Pa.), 51(4), 195-198 (2013-04-06)
Jeannette Ø Penny et al.
Acta orthopaedica, 84(2), 130-137 (2013-04-20)
Wear particles from metal-on-metal arthroplasties are under suspicion for adverse effects both locally and systemically, and the DePuy ASR Hip Resurfacing System (RHA) has above-average failure rates. We compared lymphocyte counts in RHA and total hip arthroplasty (THA) and investigated
Takuya Kurahashi et al.
Inorganic chemistry, 52(7), 3908-3919 (2013-03-23)
The Co(salen)(X) complex, where salen is chiral N,N'-bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine and X is an external axial ligand, has been widely utilized as a versatile catalyst. The Co(salen)(X) complex is a stable solid that has been conventionally described as a Co(III)(salen)(X) complex. Recent
Chuanyu Sun et al.
Journal of nanoscience and nanotechnology, 13(1), 236-241 (2013-05-08)
Owing to their great potentialities of carbon nanotubes (CNTs)-based magnetic nano-composites, numerous applications of them have been found in nanotechnology, integrated functional system, and in medicine. Herein, nearly monodisperse CoFe2O4 nanoparticles have been deposited on multi-walled carbon nanotubes (MWCNTs) by

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service