Skip to Content
Merck
All Photos(1)

Key Documents

266671

Sigma-Aldrich

Cobalt

foil, thickness 0.25 mm, ≥99.99% trace metals basis

Synonym(s):

Cobalt element, Cobalt-59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

foil

resistivity

6.24 μΩ-cm, 20°C

thickness

0.25 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

1.4 g = 25 × 25 mm; 5.6 g = 50 × 50 mm

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Perconti et al.
Journal of biological regulators and homeostatic agents, 27(2), 443-454 (2013-07-09)
Size-dependent characteristics of novel engineered nanomaterials might result in unforeseen biological responses and toxicity. To address this issue, we used cDNA microarray analysis (13443 genes) coupled with bioinformatics and functional gene annotation studies to investigate the transcriptional profiles of Balb/3T3
H John Cooper et al.
The Journal of bone and joint surgery. American volume, 95(10), 865-872 (2013-05-17)
Femoral stems with dual-taper modularity were introduced to allow additional options for hip-center restoration independent of femoral fixation in total hip arthroplasty. Despite the increasing availability and use of these femoral stems, concerns exist about potential complications arising from the
Douglas A Weigent
Cellular immunology, 282(1), 9-16 (2013-05-04)
We report here that culture of lymphoid cells under hypoxic conditions showed an increase in both luciferase expression from a GH-promoter luciferase construct and the levels of lymphocyte GH. The effect was mimicked by treatment of cells with cobalt chloride
D W Su et al.
Journal of nanoscience and nanotechnology, 13(5), 3354-3359 (2013-07-19)
Highly ordered mesoporous Co3O4 nanostructures were prepared using SBA-15 silica as hard templates. The mesoporous structures were characterized by X-ray diffraction, high resolution transmission electron microscopy, and N2 adsorption/desorption isotherm analysis. The results demonstrated that the as-prepared mesoporous Co3O4 has
Pushya A Potnis et al.
Cellular immunology, 282(1), 53-65 (2013-05-18)
Metal orthopedic implant debris-induced osteolysis of hip bone is a major problem in patients with prosthetic-hips. Although macrophages are the principal targets for implant-wear debris, the receptor(s) and mechanisms underlying these responses are not fully elucidated. We examined whether the

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service