404845
Triethylaluminum solution
1.0 M in heptane
Synonym(s):
Aluminumtriethanide
About This Item
Recommended Products
form
liquid
Quality Level
concentration
1.0 M in heptane
density
0.703 g/mL at 25 °C
SMILES string
CC[Al](CC)CC
InChI
1S/3C2H5.Al/c3*1-2;/h3*1H2,2H3;
InChI key
VOITXYVAKOUIBA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Self-heat. 2 - Skin Corr. 1A - STOT SE 3 - Water-react 1
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 2
Flash Point(F)
24.8 °F - closed cup
Flash Point(C)
-4 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service