Skip to Content
Merck
All Photos(1)

Key Documents

88144

Supelco

Tetratetracontane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)42CH3
CAS Number:
Molecular Weight:
619.19
Beilstein:
1804576
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

85-87 °C (lit.)
85-88 °C

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C44H90/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-43-44-42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-44H2,1-2H3

InChI key

KMXFZRSJMDYPPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Glaeser et al.
Ultramicroscopy, 111(2), 90-100 (2010-12-28)
Quantitative analysis of electron microscope images of organic and biological two-dimensional crystals has previously shown that the absolute contrast reached only a fraction of that expected theoretically from the electron diffraction amplitudes. The accepted explanation for this is that irradiation
Martin Göllner et al.
Advanced materials (Deerfield Beach, Fla.), 22(39), 4350-4354 (2010-08-25)
The encapsulation of pentacene thin-film transistors (TFTs) by a 50 nm thick layer of a long chain alkane (C₄₄H₉₀) enables the operation in an aqueous solution of 1 mM NaCl. It is the first time that pentacene TFTs operate successfully
F Zemlin et al.
Science (New York, N.Y.), 229(4712), 461-462 (1985-08-02)
A liquid helium-cooled cryoelectron microscope, operated to expose the specimen to only a very low electron dose, was used to obtain structural images of monolamellar n-tetratetracontane (n-C44H90) crystals at 0.25-nanometer resolution. These results are in contrast to earlier predictions that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service