262315
N-Methyl-1-naphthalenemethylamine hydrochloride
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C10H7CH2NHCH3 · HCl
CAS Number:
Molecular Weight:
207.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
191-193 °C (lit.)
functional group
amine
SMILES string
Cl.CNCc1cccc2ccccc12
InChI
1S/C12H13N.ClH/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11;/h2-8,13H,9H2,1H3;1H
InChI key
BVJVHPKFDIYQOU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
N-Methyl-1-naphthalenemethylamine hydrochloride has been used as reagent for the determination of isocyanates in air by UV or fluorescence detection. It has been used in the preparation of key intermediate required for the synthesis of terbinafine.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A New Route to the Synthesis of Terbinafine.
Lee SJ, et al.
Bull. Korean Chem. Soc., 18(11), 1218-1220 (1997)
Analytical Chemistry, 53, 1122-1122 (1981)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service