Skip to Content
Merck
All Photos(3)

Key Documents

262021

Sigma-Aldrich

Di-tert-butylsilyl bis(trifluoromethanesulfonate)

97%

Synonym(s):

DTBS ditriflate, Di-tert-butylsilyl ditriflate, Trifluoromethanesulfonic acid di-tert-butylsilylene ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)2Si[C(CH3)3]2
CAS Number:
Molecular Weight:
440.45
Beilstein:
3569211
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

73-75 °C/0.35 mmHg (lit.)

density

1.352 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](OS(=O)(=O)C(F)(F)F)(OS(=O)(=O)C(F)(F)F)C(C)(C)C

InChI

1S/C10H18F6O6S2Si/c1-7(2,3)25(8(4,5)6,21-23(17,18)9(11,12)13)22-24(19,20)10(14,15)16/h1-6H3

InChI key

HUHKPYLEVGCJTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di-tert-butylsilyl bis(trifluoromethanesulfonate), also known as di-tert-butylsilyl ditriflate, that easily reacts with hydroxylic solvents. It is commonly used as a protecting group for diols in oligonucleotide synthesis.

Application

Di-tert-butylsilyl bis(trifluoromethanesulfonate) is used:
  • As a Boekelheide promoter to synthesize pyrrolidines and piperidines via intramolecular cyclization.
  • As a protecting group reagent for 1,3-diols used recently in a synthesis of N-homoceramides.
  • For selective α-galactosylation in the synthesis of α-galactosyl ceramides.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemische Berichte, 125, 47-47 (1992)
Tetrahedron Letters, 23, 4871-4871 (1982)
Zeitschrift fur Anorganische und Allgemeine Chemie, 618, 144-144 (1992)
Synlett, 2379-2379 (2006)
A new sequential intramolecular cyclization based on the boekelheide rearrangement
Assunta M,et al.
European Journal of Organic Chemistry (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service