Skip to Content
Merck
All Photos(4)

Key Documents

226149

Sigma-Aldrich

tert-Butyldimethylsilyl trifluoromethanesulfonate

reagent grade, 98%

Synonym(s):

TBDMS triflate, Trifluoromethanesulfonic acid tert-butyldimethylsilylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
264.34
Beilstein:
2370068
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

98%

refractive index

n20/D 1.385 (lit.)

bp

65-67 °C/12 mmHg (lit.)

density

1.151 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

InChI

1S/C7H15F3O3SSi/c1-6(2,3)15(4,5)13-14(11,12)7(8,9)10/h1-5H3

InChI key

WLLIXJBWWFGEHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This reagent was used to introduce a bulky tert-butyl dimethylsilyl group onto a cis-bis(alkenyl)oxirane facilitating a room temperature Cope rearrangement to a 2-CF3-4,5-dihydrooxepin.
Together with a thiolane promotes a rapid and efficient chalcogenide-Morita-Baylis-Hillman reaction.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 625-625 (1993)
Journal of the Chemical Society. Chemical Communications, 1280-1280 (1988)
Chemistry Letters (Jpn), 33, 438-439 (2004)
Tetrahedron Letters, 47, 3553-3553 (2006)
The Journal of Organic Chemistry, 58, 2287-2287 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service