Skip to Content
Merck
All Photos(1)

Key Documents

253081

Sigma-Aldrich

Titanium(IV) propoxide

98%

Synonym(s):

Tetrapropyl orthotitanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti(OC3H7)4
CAS Number:
Molecular Weight:
284.22
Beilstein:
4146685
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid
liquid

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.499 (lit.)

bp

170 °C/3 mmHg (lit.)

density

1.033 g/mL at 25 °C (lit.)

SMILES string

CCCO[Ti](OCCC)(OCCC)OCCC

InChI

1S/4C3H7O.Ti/c4*1-2-3-4;/h4*2-3H2,1H3;/q4*-1;+4

InChI key

HKJYVRJHDIPMQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher P Nicholas et al.
Langmuir : the ACS journal of surfaces and colloids, 20(22), 9456-9462 (2004-10-20)
The sulfated metal oxides (SMOs) sulfated stannia (SnS), sulfated iron oxide (FeS), and sulfated titanium dioxide (TiS) have been synthesized and examined as support materials/cocatalysts/activators for molecule-based olefin polymerization and hydrogenation catalysis. (13)C CPMAS NMR spectroscopic analysis of Cp(2)Zr((13)CH(3))(2)/SMO chemisorption
Jyun-Hong Shen et al.
Chemosphere, 251, 126380-126380 (2020-03-13)
A detailed mechanistic investigation of the hydroxyl radical (•OH) formation and organic pollutant degradation over transition metal-doped and undoped TiO2 photocatalysts was performed by the quantitative measurement of •OH and the identification of intermediate products under various experimental conditions. The
Dominique Mombrú et al.
Scientific reports, 9(1), 5322-5322 (2019-03-31)
Here, we report large magnetoresistance and magnetocapacitance response of undoped TiO2 quantum dots weighting the contribution of both grain and grain boundaries by means of impedance spectroscopy. We also performed a complete characterization of the TiO2 quantum dots (~5 nm) prepared
Yasutaka Kuwahara et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(2), 380-389 (2016-11-26)
One-pot reaction involving Pd-catalyzed H
Donghee Kim et al.
Nanoscale, 8(13), 7000-7016 (2015-11-03)
Despite a large number of developments of noble metal (or metal oxide) NP-based catalysts, it has been a great challenge to prepare high-performance recyclable catalysts with integrated functionalities that can be used in various solvent media. Here, we report on

Articles

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service