Skip to Content
Merck
All Photos(2)

Key Documents

252875

Sigma-Aldrich

N,N-Dimethylpropionamide

98%

Synonym(s):

DMP, N,N-Di-methyl-3-propanamide, N,N-Dimethylpropanamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CON(CH3)2
CAS Number:
Molecular Weight:
101.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

174-175 °C (lit.)

mp

−45 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

CCC(=O)N(C)C

InChI

1S/C5H11NO/c1-4-5(7)6(2)3/h4H2,1-3H3

InChI key

MBHINSULENHCMF-UHFFFAOYSA-N

General description

Solvation structure of the zinc (II) ion in N,N-dimethylpropionamide was studied by Raman spectroscopy. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

Application

N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiang Ming et al.
Nanoscale, 12(6), 3916-3930 (2020-02-01)
Palladium nanosheets (Pd NSs) have recently attracted increasing research interest in the biomedical field due to their excellent near-infrared absorption, photothermal conversion capability and biocompatibility. However, the application of Pd NSs in immunotherapy has not been reported. Here, Pd NSs
Yasuhiro Umebayashi et al.
The journal of physical chemistry. A, 109(21), 4862-4868 (2006-07-13)
Solvation structure of the zinc(II) ion in N,N-dimethylpropionamide (DMPA) was studied by Raman spectroscopy at varying temperature and by quantum mechanical calculations. No significant ion-pair formation was found for the Zn(ClO4)2 solution in the molality range m(Zn) < 1.5 mol
Saige Shi et al.
ACS applied materials & interfaces, 7(26), 14369-14375 (2015-06-16)
Palladium nanosheets with strong near-infrared absorption have been recently demonstrated as promising photothermal agents for photothermal therapy (PTT) of cancers. However, systematic assessments of their potential risks and impacts to biological systems have not been fully explored yet. In this
Solvation structure and complexation of the manganese (II) ion in N, N-dimethylpropionamide and N, N, N', N'-tetramethylurea studied by means of titration calorimetry and Raman spectroscopy.
Zhang Y, et al.
Journal of Solution Chemistry, 34(12), 1429-1443 (2005)
Daniel Lundberg et al.
Physical chemistry chemical physics : PCCP, 20(21), 14525-14536 (2018-05-17)
The structures of the N,N-dimethylformamide (dmf), N,N-dimethylacetamide (dma), and N,N-dimethylpropionamide (dmp) solvated strontium and barium ions have been determined in solution using large angle X-ray scattering and EXAFS spectroscopy. The strontium ion has a mean coordination number (CN) between 6.2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service