Skip to Content
Merck
All Photos(1)

Key Documents

146358

Sigma-Aldrich

1-Ethyl-2-pyrrolidone

98%

Synonym(s):

NEP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
107971
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

97 °C/20 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

SMILES string

CCN1CCCC1=O

InChI

1S/C6H11NO/c1-2-7-5-3-4-6(7)8/h2-5H2,1H3

InChI key

ZFPGARUNNKGOBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-2-pyrrolidone is a transdermal absorption-enhancing compound and mechanism of its effect on multilammellar liposome of stratum corneum lipid has been studied.

Application

1-Ethyl-2-pyrrolidone was used to modify coir fiber (from Cocos nucifera) by photocuring.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Pretreatment with Ultraviolet Radiation on the Physicomechanical Properties of Photocured Coir (Cocos nucifera) Fiber with 1-Ethyl-2-Pyrrolidone (1-E-2-P) Monomer.
Khan MA, et al.
Polymer-Plastics Technology and Engineering, 45(4), 487-494 (2006)
Effect of penetration enhancers (pyrrolidone derivatives) on multilamellar liposomes of stratum corneum lipid: a study by UV spectroscopy and differential scanning calorimetry.
Kim C-K, et al.
International Journal of Pharmaceutics, 95(1), 43-50 (1993)
Ajeet Kaushik et al.
Scientific reports, 7, 45663-45663 (2017-04-05)
In this research, we demonstrate cell uptake of magneto-electric nanoparticles (MENPs) through nanoelectroporation (NEP) using alternating current (ac)-magnetic field stimulation. Uptake of MENPs was confirmed using focused-ion-beam assisted transmission electron microscopy (FIB-TEM) and validated by a numerical simulation model. The
Takeshi Katsuda et al.
Scientific reports, 3, 1197-1197 (2013-02-05)
Alzheimer's disease (AD) is characterized by the accumulation of β-amyloid peptide (Aβ) in the brain because of an imbalance between Aβ production and clearance. Neprilysin (NEP) is the most important Aβ-degrading enzyme in the brain. Thus, researchers have explored virus-mediated
Xiaolei Zhu et al.
Aging cell, 16(5), 1073-1082 (2017-08-05)
The accumulation and deposition of beta-amyloid (Aβ) is a key neuropathological hallmark of Alzheimer's disease (AD). Histone deacetylases (HDACs) are promising therapeutic targets for the treatment of AD, while the specific HDAC isoforms associated with cognitive improvement are poorly understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service