Skip to Content
Merck
All Photos(2)

Key Documents

250988

Sigma-Aldrich

2,5-Dibromotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3Br2
CAS Number:
Molecular Weight:
249.93
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.602 (lit.)

bp

135-136 °C/35 mmHg (lit.)

mp

5-6 °C (lit.)

density

1.815 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Cc1cc(Br)ccc1Br

InChI

1S/C7H6Br2/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3

InChI key

QKEZTJYRBHOKHH-UHFFFAOYSA-N

Application

2,5-Dibromotoluene has been used:
  • as internal standard for quantification of cyanide and thiocyanate in human saliva by GC-MS
  • in the preparation of 2,5-bis(4′-alkoxycarbonylphenyl)styrenes, monomers required for the synthesis of mesogen-jacketed liquid-crystal polymers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buddha D Paul et al.
Journal of analytical toxicology, 30(8), 511-515 (2006-11-30)
A method is described for simultaneous determination of cyanide (CN) and thiocyanate (SCN) in human saliva, or oral fluid. SCN concentrations in body fluids appeared to be important in classifying patients as smokers or nonsmokers, in determining some clinical conditions
Synthesis and characterization of mesogen-jacketed liquid-crystal polymers based on 2, 5-bis (4'-alkoxyphenyl) styrene.
Yu Z, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 41(10), 1454-1464 (2003)
Ekaterina A Dolgopolova et al.
Journal of the American Chemical Society, 141(13), 5350-5358 (2019-03-07)
Electronic structure modulation of metal-organic frameworks (MOFs) through the connection of linker "wires" as a function of an external stimulus is reported for the first time. The established correlation between MOF electronic properties and photoisomerization kinetics as well as changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service